Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Standard 50ppm/H2O, Reagents

Catalog No. p-200021842
Click to view available options
Quantity:
1 L

Calcium Standard 50ppm/H2O, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Calcium Standard 50ppm/H2O
Packaging Poly Bottle
CAS 7732-18-5, 10035-04-8
Quantity 1 L
Specific Gravity 1.0003
For Use With (Application) Water Analysis
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.