Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO3, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Plastic bottle
7440-70-2,7697-37-2,7732-18-5
Ca
40.078
MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
5460341
CHEBI:29320
calcium
[Ca]
7440-70-2,7697-37-2,7732-18-5
Ca
40.078
MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
5460341
CHEBI:29320
calcium
[Ca]
for atomic adsorption spectroscopy
Concentration 997 to 1003 μg/mL of Ca(2+) in 0.5N HNO3 (20°C)
Formula Weight 40.08
Physical Form Solution
Grade AAS
Packaging Plastic bottle
Solubility Solubility in water: soluble.
Color Undesignated
Quantity 500mL
Chemical Name or Material Calcium standard solution, 1 mg/ml Ca in 0.5N HNO3

GHS H Statement:
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May be corrosive to metals.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger