Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Stearate, NF, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1592-23-0
C36H70CaO4
607.03
CJZGTCYPCWQAJB-UHFFFAOYSA-L
calcium dioctadecanoate
[Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
1592-23-0
C36H70CaO4
607.03
CJZGTCYPCWQAJB-UHFFFAOYSA-L
calcium dioctadecanoate
[Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

CA169, 1592-23-0

Spectrum™ Calcium Stearate, NF has good anti-adherent and lubricant properties and is used for tablet and capsule lubrication. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Loss on Drying 4%
Packaging Fiber Drum
Quantity 45 kg