Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Calcium Sulfate, 99%, −325 mesh, Honeywell™ Fluka™

Click to view available options
Quantity:
100g
500g
Packaging:
Poly Bottle
7778-18-9
CaO4S
136.134
MFCD00010912
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium sulfate,drierite,gypsum,sulfuric acid, calcium salt 1:1,karstenite,calcium sulphate,anhydrous gypsum,plaster of paris,anhydrous calcium sulfate,calcium sulfate, anhydrous
24497
CHEBI:31346
calcium;sulfate
[O-]S(=O)(=O)[O-].[Ca+2]
7778-18-9
CaO4S
136.134
MFCD00010912
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium sulfate,drierite,gypsum,sulfuric acid, calcium salt 1:1,karstenite,calcium sulphate,anhydrous gypsum,plaster of paris,anhydrous calcium sulfate,calcium sulfate, anhydrous
24497
CHEBI:31346
calcium;sulfate
[O-]S(=O)(=O)[O-].[Ca+2]

−325 mesh, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula CaSO4
UN Number NONH for all modes of transport
Appearance Powder
Formula Weight 136.14g/mol
Percent Purity 99%
Density 2.960g/cm3
Mesh Size −325mesh
Packaging Poly Bottle
Color White
Quantity 500g
Chemical Name or Material Calcium Sulfate
Show More Show Less

EINECSNumber : 231-900-3

RTECSNumber : WS6920000