Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Sulfate, Anhydrous, FCC, 98%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7778-18-9
CaO4S
136.13
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium sulfate
[Ca++].[O-]S([O-])(=O)=O
7778-18-9
CaO4S
136.13
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium sulfate
[Ca++].[O-]S([O-])(=O)=O

CA171, 7778-18-9, CaSO4

Spectrum™ Calcium Sulfate, Anhydrous, FCC is used in the food industry as an anticaking agent, dough strengthener and stabilizer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Lead (Pb) 2 mg/kg
Percent Purity 98%
Loss on Drying 1.5%
Packaging Poly Bottle
Quantity 500 g