Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Sulfate, Anhydrous, Powder, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
7778-18-9
7778-18-9
Grade Reagent
CAS Max % 1
Chloride 0.0001
Heavy Metals 3e-05
Nitrate 5e-05
Packaging Poly Bottle
Quantity 125 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.