Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium sulfate dihydrate, 98+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic bottle
10101-41-4
CaO42H2O
172.17
PASHVRUKOFIRIK-UHFFFAOYSA-L
calcium sulfate dihydrate,phosphogypsum,landplaster,annaline,compactrol,gips,gypsite,primoplast,satinite,gypsum stone
24928
CHEBI:32583
calcium;sulfate;dihydrate
O.O.[O-]S(=O)(=O)[O-].[Ca+2]
10101-41-4
CaO42H2O
172.17
PASHVRUKOFIRIK-UHFFFAOYSA-L
calcium sulfate dihydrate,phosphogypsum,landplaster,annaline,compactrol,gips,gypsite,primoplast,satinite,gypsum stone
24928
CHEBI:32583
calcium;sulfate;dihydrate
O.O.[O-]S(=O)(=O)[O-].[Ca+2]
Assay Percent Range 98% min. (Complexometry)
Linear Formula CaSO4·2H2O
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: slightly soluble in glycerol,practically insoluble in most common organic,solvents
Formula Weight 172.17
Percent Purity 98+%
Grade Extra Pure
Packaging Plastic bottle
Solubility Solubility in water: 2g/L (20°C). Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents
Quantity 5kg
Chemical Name or Material Calcium sulfate dihydrate