Alfa Aesar™ Calcium sulfate dihydrate, 99%

Manufacturer: Alfa Aesar™ 33301A1

Quantity 1kg

 View more versions of this product

Catalog No. AA33301A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10101-41-4
    Molecular Formula CaSO4·2H2O
    Formula Weight 172.17 (136.14 Anhydrous)
    MDL Number MFCD00149625
    Merck Index 14,1706
    Chemical Name or Material Calcium sulfate dihydrate

    Specifications

    Formula Weight 172.17 (136.14 Anhydrous)
    Boiling Point 163°C (in - 2H2O)
    Melting Point 128°C (in -1.5H2O)
    Assay Percent Range 99%
    Molecular Formula CaSO4·2H2O
    Odor Odorless
    Sensitivity Hygroscopic
    Density 2.32g/mL
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00149625
    Merck Index 14,1706
    Quantity 1kg
    RTECS Number EW4150000
    CAS 10101-41-4
    Chemical Name or Material Calcium sulfate dihydrate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit