Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Sulfate, Dihydrate, ACS, 98-102%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10101-41-4
10101-41-4
Grade ACS
CAS Max % 1
Percent Purity 98 to 102%
Chloride 5e-05
Heavy Metals 2e-05
Iron (Fe) 1e-05
Magnesium (Mg) 0.0002
Packaging Poly Bottle
Potassium (K) 5e-05
Sodium (Na) 0.0002
Strontium (Sr) 0.0005
Quantity 500 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.