Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium sulfate hemihydrate, 97+%, pure, Acros Organics

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
10034-76-1
0·5 H2O
145.15
MFCD00149624
ZOMBKNNSYQHRCA-UHFFFAOYSA-J
calcium sulfate hemihydrate,gypsum hemihydrate,calcium sulphate hemihydrate,unii-3rw091j48v,crystacal,densite,hemihydrate gypsum,densite gypsum,gipshalbhydrat,p 23 sulfate
3033839
CHEBI:32584
dicalcium;disulfate;hydrate
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ca+2].[Ca+2]
10034-76-1
0·5 H2O
145.15
MFCD00149624
ZOMBKNNSYQHRCA-UHFFFAOYSA-J
calcium sulfate hemihydrate,gypsum hemihydrate,calcium sulphate hemihydrate,unii-3rw091j48v,crystacal,densite,hemihydrate gypsum,densite gypsum,gipshalbhydrat,p 23 sulfate
3033839
CHEBI:32584
dicalcium;disulfate;hydrate
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ca+2].[Ca+2]
Assay Percent Range 97+%
Solubility Information Solubility in water: 0.8g/L (20°C)
Formula Weight 145.15
Physical Form Powder
Percent Purity ≥97%
Grade Pure
Identification Confirmed
Loss on Ignition 4.5% to 8.0% (500°C, 2 Hrs)
Packaging Plastic bottle
Solubility Solubility in water: 0.8g/L (20°C).
Color White
Quantity 100g
Chemical Name or Material Calcium sulfate hemihydrate
Show More Show Less