Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Sulfate Solution (Saturated), Reagents
Click to view available options
Quantity: 10 L
Specifications
Specifications
Boiling Point | 100°C |
Chemical Name or Material | Calcium Sulfate Solution (Saturated) |
Color | Colorless |
CAS | 7732-18-5, 10101-41-4 |
Physical Form | Liquid |
Quantity | 10 L |
Grade | Reagent |
Packaging | Cubitainer |
Specific Gravity | 1.0019 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction