Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Sulfate TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
Poly Bottle
7778-18-9,7732-18-5
CaO4S
136.13
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium sulfate
[Ca++].[O-]S([O-])(=O)=O
7778-18-9,7732-18-5
CaO4S
136.13
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium sulfate
[Ca++].[O-]S([O-])(=O)=O

C-328, 7778-18-9

Spectrum™ Calcium Sulfate TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS Max % 0.1%,99.9%
Packaging Amber Glass Bottle
Quantity 4 L