Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium trifluoromethanesulfonate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
55120-75-7
C2CaF6O6S2
338.204
MFCD00143911
PUQLFUHLKNBKQQ-UHFFFAOYSA-L
calcium trifluoromethanesulfonate,calcium triflate,methanesulfonic acid, trifluoro-, calcium salt,calcium trifluoromethansulphonate,methanesulfonic acid, 1,1,1-trifluoro-, calcium salt 2:1,calcium ditriflate,calcium trifluoromethyl sulfonate,calcium ii triflate,calcium 2+ ditriflate
2734307
calcium;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ca+2]
55120-75-7
C2CaF6O6S2
338.204
MFCD00143911
PUQLFUHLKNBKQQ-UHFFFAOYSA-L
calcium trifluoromethanesulfonate,calcium triflate,methanesulfonic acid, trifluoro-, calcium salt,calcium trifluoromethansulphonate,methanesulfonic acid, 1,1,1-trifluoro-, calcium salt 2:1,calcium ditriflate,calcium trifluoromethyl sulfonate,calcium ii triflate,calcium 2+ ditriflate
2734307
calcium;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ca+2]
Quantity 25g
Color Gray
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula (F3CSO3)2Ca
Formula Weight 338.21
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material Calcium trifluoromethanesulfonate, 98%

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.