Alfa Aesar™ Calcium zirconium isopropoxide, 10% w/v in isopropanol

Precursor for mixed Ca, Zr oxide by Sol-Gel method

Manufacturer: Alfa Aesar™ 41971AE

Quantity 100mL

 View more versions of this product

Catalog No. AA41971AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 334512-82-2
    Linear Formula CaZr[OCH(CH3)2]6
    Formula Weight 485.83
    MDL Number MFCD01865977
    UN Number UN1219
    Chemical Name or Material Calcium zirconium isopropoxide

    Specifications

    Formula Weight 485.83
    Health Hazard 3 GHS P Statement
    P210-P280g-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    MDL Number MFCD01865977
    Concentration 10% w/v in isopropanol
    Physical Form Liquid
    UN Number UN1219
    Color Yellow
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    Linear Formula CaZr[OCH(CH3)2]6
    CAS 334512-82-2
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H336
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    Quantity 100mL
    Sensitivity Moisture Sensitive
    Chemical Name or Material Calcium zirconium isopropoxide

    GHS H Statement
    H225-H315-H319-H336
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P280g-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit