Alfa Aesar™ Calcium zirconium oxide, 99.2% (metals basis)

Manufacturer: Alfa Aesar™ 1234430

Quantity 250g

 View more versions of this product

Catalog No. AA1234430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12013-47-7
    Formula Weight 179.3
    MDL Number MFCD00015982
    Synonym Calcium zirconate
    Chemical Name or Material Calcium zirconium oxide

    Specifications

    Formula Weight 179.3
    Synonym Calcium zirconate
    Melting Point 2550°C
    TSCA TSCA
    Mesh Size −325 Mesh
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00015982
    CAS 12013-47-7
    Quantity 250g
    Chemical Name or Material Calcium zirconium oxide
    Assay 99.2%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit