Learn More
Abnova™ CALCOCO1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00057658T01
Catalog No. 89-937-838
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CALCOCO1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 77.3kDa
Transient overexpression cell lysate was tested with anti-CALCOCO1 antibody (Mfr. No. H00057658-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_020898
pCMV-CALCOCO1 full-length
100 μL
Human