Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caldesmon/CALD1 Mouse anti-Human, Biotin, Clone: CALD1/820 + h-CALD, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP247819B

Catalog No. NBP47819B


Add to cart

Description

Description

Caldesmon/CALD1 Monoclonal antibody specifically detects Caldesmon/CALD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications

Specifications

Caldesmon/CALD1
CALD1/820 + h-CALD
Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Purified
CALD1
Recombinant full-length human CALD1 protein (CALD1/820); Crude human uterus extract (h-CALD)
Protein A or G purified
Cytoskeleton Markers, Phospho Specific
Primary
800
Human
Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Biotin
PBS with 0.05% Sodium Azide
CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22
Mouse
IgG
0.1 ml
Store at 4C in the dark.
Monoclonal
Recognizes a protein of 150kDa, which is identified as the high molecular weight variant of Caldesmon. Two closely related variants of human caldesmon have been identified which are different in their electrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70-80kDa) is found in non- muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle. This MAb recognizes only the 150kDa variant (h-caldesmon) in Western blots of human aortic media extracts and is unreactive with fibroblast extracts from cultivated human foreskin. Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit