Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caldesmon/CALD1 Mouse anti-Human, Clone: rCALD1/820, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen Caldesmon/CALD1
Clone rCALD1/820
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122208
View Documents
Novus Biologicals
NBP30736720UG
20 μg
Each for $159.82
Add to cart
 
NB122207
View Documents
Novus Biologicals
NBP30736710UG
100 μg
Each for $374.34
Add to cart
 
Description

Description

Caldesmon/CALD1 Monoclonal antibody specifically detects Caldesmon/CALD1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

Caldesmon/CALD1
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin)
Purified
RUO
Human
CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22
Recombinant full-length human Caldesmon/CALD1 protein
Monoclonal
Protein A or G purified
rCALD1/820
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml
Unconjugated
Mouse
Cell Biology, Cellular Markers, Phospho Specific
10 mM PBS with 0.05% BSA
800
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit