Learn More
Caldesmon, HMW Mouse anti-Human, Clone: SPM168, Abnova™
Mouse monoclonal antibody raised against native human Caldesmon, HMW.
Manufacturer:  Abnova Corporation MAB13178
Catalog No. 89-933-711
Add to cart
Description
Description
This gene encodes a calmodulin- and actin-binding protein that plays an essential role in the regulation of smooth muscle and nonmuscle contraction. The conserved domain of this protein possesses the binding activities to Ca(2+)-calmodulin, actin, tropomyosin, myosin, and phospholipids. This protein is a potent inhibitor of the actin-tropomyosin activated myosin MgATPase, and serves as a mediating factor for Ca(2+)-dependent inhibition of smooth muscle contraction. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]
Specifications
Specifications
Caldesmon, HMW
SPM168
Mouse monoclonal antibody raised against native human Caldesmon, HMW.
In 10mM PBS.
CDM/H-CAD/L-CAD/MGC21352/NAG22
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
CALD1
Crude human uterus extract.
Protein A/G purification
RUO
Primary
800