Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dukal™ Caliber™ Type VII Gauze Sponges
These Type VII Gauze Sponges are cotton, highly absorbent and offer exceptional clinical performance.
Supplier: Dukal™ C82122
This item is not returnable.
View return policy
Description
- Meets USP Type VII requirements.
- 100% woven cotton.
- Highly absorbent and offers exceptional clinical performance.
- Used for a number of applications including: general wound care, dressings, prepping, wound packing, and debriding.
Specifications
Non-sterile | |
2 x 2 in. | |
Yes | |
12-Ply |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction