Learn More
Thermo Scientific Water Analysis Instrument Calibration and Repair Service
Thermo Scientific provides calibration and repair services for electrochemistry devices for water analysis including pH meters. Instruments are issued calibration certificates that are traceable to NIST standards and can be used for internal compliance and audit.
Manufacturer: Thermo Scientific CALSERVD2
Description
Out of tolerance instruments can be adjusted or repaired as needed. Thermo Scientific also offers cost-effective device exchanges and loaner services, as well as expedited services for devices that need to be repaired within 24 - 48 hours.
Benefits:
- Helps meet ISO, FDA, USDA, EPA, GLP/cGMP quality assurance guidelines before use in critical processes
- Instruments are calibrated to factory standards and include before (as found) and after (as left) data to
document instrument accuracy - Gain confidence in measurement precision
- Traceability chain for the standards used in calibration
- Protect your investment and prevent equipment downtime
- pH calibration service includes measurement of meter accuracy at five test points across pH range and five test points across corresponding mV range
- Calibration services performed by highly skilled technicians and backed by a 90-day satisfaction guarantee
To help meet regulatory requirements, all Thermo Scientific Orion Water Analysis Instrument Test Equipment is traceable to NIST.
Certification of Thermo Scientific's test equipment is compliant with ANSI/NCSL ZA540-1 standards, ISO 9001:2015, and ISO 9001:2005 requirements.
Annual calibrations from the date your equipment is put into operation are recommended.
Thermo Scientific's calibration services specialize in servicing the following Thermo Scientific Orion Water Analysis Instrumentation:
- Thermo Scientific Orion Versa Star Pro
- Thermo Scientific Orion Dual Star
- Thermo Scientific Orion Star A
- Thermo Scientific Orion Star
- Thermo Scientific Orion Star T900 Series
- Thermo Scientific Orion Conductivity Resistor Kits
Specifications
Thermo Scientific | |
Calibration Level 2 | |
Calibration and repair services for electrochemistry devices for water analysis including pH meters | |
Orion: Star A215, Star A216, Star A325, Star A326, Versa Star (2 Modules), 4-Star/4-Star Plus; Accumet: AB200, AB250, XL200, XL250, AP84, AP85, AP125 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.