Learn More
Calibrators for Roche Diagnostics cobas™ c 111 Chemistry Analyzer
Manufacturer:  Roche Diagnostics 11447394216
 View more versions of this product
Catalog No. 22-045-816
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CFAS CK-MB Calibrator
Roche Cobas™ c 111 chemistry analyzer
3 x 1mL
Human Serum, Pentaerythrite, HEPES, Tetramethylammonium Chloride, Sodium Chloride, Glucose
Keep containers dry and tightly closed to avoid moisture absorption and contamination