Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Calibrex™ organo 525 Bottle Top Dispensers

Adjustable dispensers include a ground glass plunger that is best suited for organic, non-crystallizing acid and base solutions.

Manufacturer:  DWK Life Sciences W845000

 View more versions of this product

Catalog No. 03-410-900


  Check Availability
Add to cart

Description

Description

  • Permanent fluid path visibility
  • Integrated calibration system
  • Choice of adjustment button with each unit
  • Long lasting performance stability
  • Autoclavable at 121°C / 250°F fully assembled
  • Two year warranty
Specifications

Specifications

0.1–1mL
Analog
Calibrex organo 525
Yes
Bottle Top Dispenser
2 Years
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit