Learn More
Aat BioquestSupplier Diversity Partner California Red™ SE DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 479
Although Texas Red® is the most popular labeling reagent of sulfonyl chloride, it is quite unstable in water. Even in anhydrous DMF, Texas Red® tends to give a very complicated reaction mixture.
Catalog No. 50-195-2896