Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALML3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179929

 View more versions of this product

Catalog No. NBP179929


Add to cart

Description

Description

CALML3 Polyclonal antibody specifically detects CALML3 in Human, Rat, Porcine, Yeast samples. It is validated for Western Blot.
Specifications

Specifications

CALML3
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
CALML3
The immunogen for this antibody is CALML3. Peptide sequence TVDFPEFLGMMARKMKDTDNEEEIREAFRVFDKDGNGFVSAAELRHVMTR.
16 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Pig: 92%.
Human, Porcine, Rat, Yeast
Western Blot
Western Blot 1:1000
NP_005176
calmodulin-like 3, calmodulin-like protein 3, CaM-like protein, CLPCalmodulin-related protein NB-1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
810
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit