Learn More
CALML4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158941AP
Catalog No. 50-173-0557
Add to cart
Specifications
Specifications
CALML4
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91WQ9, Q96GE6
CALML4, calmodulin like 4, Calmodulin like protein 4, NY BR 20
Rabbit
IgG
150 μL
-20°C
Primary
691455, 75600, 91860
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
CALML4
Liquid
Calml4
CALML4 Fusion Protein Ag8705
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat