Sino Biological™ CALML5 Recombinant Human Protein, N-His.GST tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human CALML5 (Met 1-Glu 146) fused to N-terminal polyhistidine-tagged GST tag at N-terminus

Manufacturer: Sino Biological™ 11783H20E50

 View more versions of this product

Catalog No. 11-783-H20E50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Structural Form Full Length, Recombinant
    Molecular Weight 44.2kd
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Source Human
    Endotoxin Concentration <1 EU/μg
    Common Name Calmodulin-Like 5
    Gene Symbol CALML5
    Protein Tag GST Tag, His Tag
    Validated Application Assay Standard
    Protein Family Other proteins
    Expression System E.coli
    Product Type CALML5 Recombinant Human Protein
    For Use With (Application) Analysis of protein stucture and used in cell biology research applications
    Format Lyophilized

    Expressed from DNA sequence encoding human CALML5 (Met 1-Glu 146) fused to N-terminal polyhistidine-tagged GST tag at N-terminus

    • Formulation: 20mM Tris, 150mM NaCl, 1mM DTT, 0.5mM GSH, 10% Glycerol, pH 7.4

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit