Learn More
Calnexin Mouse anti-Human, Alexa Fluor™ 532, Clone: IE2.1C12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP236571AF532
Catalog No. NP23671A32
Add to cart
Description
Description
Calnexin Monoclonal antibody specifically detects Calnexin in Human samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Flow Cytometry.
Specifications
Specifications
Calnexin
IE2.1C12
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
CANX
Partial recombinant human Calnexin protein (between amino acids 1-300) [UniProtNo. P27824]
0.1 mL
Primary
821