Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calnexin, Mouse anti-Human, DyLight 488, Clone: CANX/1543, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279835G
Description
Calnexin Monoclonal antibody specifically detects Calnexin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Calnexin | |
CANX/1543 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
CANX | |
Recombinant N-terminal fragment of human Calnexin protein (around aa 1-300) (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision | |
Primary | |
This antibody recognizes a protein of 90kDa, which is identified as Calnexin. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title