Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: 1563, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243765V

Catalog No. NBP243765V


Add to cart

Description

Description

Calnexin Monoclonal antibody specifically detects Calnexin in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Calnexin
1563
Western Blot
Purified
CANX
Recombinant protein encompassing a sequence within the center region of human Calnexin. The exact sequence is proprietary.
Protein G purified
ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision
Primary
821
Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit