Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Mouse anti-Human, Rat (Negative), PE/Atto594, Clone: CANX/1541, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254552PEATT594

Catalog No. N25452PE594


Add to cart

Description

Description

Calnexin Monoclonal antibody specifically detects Calnexin in Human, Rat (Negative) samples. It is validated for Flow Cytometry.
Specifications

Specifications

Calnexin
CANX/1541
Flow Cytometry
Conjugated, Purified
CANX
Recombinant N-terminal fragment of human Calnexin protein (exact sequence is proprietary)
Protein A or G purified
ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision
Primary
821.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Rat (Negative)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit