Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain 1 Antibody (CAPN1/1530) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308456
Description
Calpain 1 Monoclonal specifically detects Calpain 1 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
Calpain 1 | |
Monoclonal | |
1 mg/mL | |
ELISA : 2-4 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL | |
Calcium-activated neutral proteinase 1, calpain 1, (mu/I) large subunit, Calpain mu-type, calpain, large polypeptide L1, calpain-1 catalytic subunit, Calpain-1 large subunit, CANP, CANP 1, CANPL1EC 3.4.22.52, Cell proliferation-inducing gene 30 protein, cell proliferation-inducing protein 30, EC 3.4.22, Micromolar-calpain, muCANPCANP1, muCL | |
Recombinant full-length human Calpain 1 protein | |
100 μg | |
Cell Cycle and Replication | |
823 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin) | |
CAPN1/1530 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction