Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain 11 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179487

 View more versions of this product

Catalog No. NBP179487


Add to cart

Description

Description

Calpain 11 Polyclonal antibody specifically detects Calpain 11 in Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Sheep, Zebrafish samples. It is validated for Western Blot.
Specifications

Specifications

Calpain 11
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
CAPN11
Synthetic peptide directed towards the N terminal of human CAPN11. Peptide sequence NNSRLKAKGVGQHDNAQNFGNQSFEELRAACLRKGELFEDPLFPAEPSSL.
80 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Horse: 86%; Goat: 85%; Sheep: 85%; Bovine: 85%; Mouse: 77%.
Bovine, Canine, Equine, Goat, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Western Blot
Western Blot 1:1000
NP_008989
Calcium-activated neutral proteinase 11, calpain 11, calpain11, calpain-11, CANP 11, EC 3.4.22, EC 3.4.22.-, EC 3.4.22.52, EC 3.4.22.54
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
11131
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit