Learn More
Calpain 2 Antibody (OTI1F4), FITC, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270335F
Catalog No. NBP270335F
Add to Cart
Description
Description
Calpain 2 Monoclonal specifically detects Calpain 2 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.
Specifications
Specifications
Calpain 2
Monoclonal
FITC
PBS with 0.05% Sodium Azide
CAPN2
Full length recombinant protein of human Calpain 2 (NP_001739) produced in HEK293T cells.
0.1 mL
824.0
Store at 4°C in the dark.
Flow Cytometry
OTI1F4
Flow Cytometry
Calcium-activated neutral proteinase 2, calpain 2, (m/II) large subunit, calpain 2, large [catalytic] subunit, calpain 2, large subunit, Calpain large polypeptide L2, Calpain M-type, calpain, large polypeptide L2, calpain-2 catalytic subunit, Calpain-2 large subunit, CANP 2, CANPL2CANP2, CANPml, EC 3.4.22, EC 3.4.22.53, FLJ39928, M-calpain, mCANP, Millimolar-calpain
Mouse
Affinity Purified
Primary
Human, Mouse, Rat, Monkey
IgG2a
Product Suggestions
Product Suggestions