Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain 2 Antibody (OTI3G1) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270334

Catalog No. NBP270334


Add to Cart

Description

Description

Calpain 2 Monoclonal specifically detects Calpain 2 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Calpain 2
Monoclonal
LYOPH
Western Blot 1:2000, Immunohistochemistry 1:150
Calcium-activated neutral proteinase 2, calpain 2, (m/II) large subunit, calpain 2, large [catalytic] subunit, calpain 2, large subunit, Calpain large polypeptide L2, Calpain M-type, calpain, large polypeptide L2, calpain-2 catalytic subunit, Calpain-2 large subunit, CANP 2, CANPL2CANP2, CANPml, EC 3.4.22, EC 3.4.22.53, FLJ39928, M-calpain, mCANP, Millimolar-calpain
Mouse
79.8 kDa
100 μg
824
Human, Mouse, Rat, Canine
Lyophilized
Western Blot, Immunohistochemistry
OTI3G1
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
CAPN2
Full length human recombinant protein of human CAPN2(NP_001739) produced in HEK293T cell.
Affinity Purified
Primary
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Store at -20°C. Avoid freeze-thaw cycles.
IgG2a
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit