Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain 2 Mouse anti-Human, Mouse, Rat, Monkey, Janelia Fluor™ 646, Clone: OTI1F4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270335JF646

Catalog No. N270335J646


Add to cart

Description

Description

Calpain 2 Monoclonal antibody specifically detects Calpain 2 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

Calpain 2
OTI1F4
Flow Cytometry
Affinity Purified, Conjugated
CAPN2
Full length recombinant protein of human Calpain 2 (NP_001739) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human, Monkey, Mouse, Rat
Flow Cytometry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Calcium-activated neutral proteinase 2, calpain 2, (m/II) large subunit, calpain 2, large [catalytic] subunit, calpain 2, large subunit, Calpain large polypeptide L2, Calpain M-type, calpain, large polypeptide L2, calpain-2 catalytic subunit, Calpain-2 large subunit, CANP 2, CANPL2CANP2, CANPml, EC 3.4.22, EC 3.4.22.53, FLJ39928, M-calpain, mCANP, Millimolar-calpain
Mouse
IgG2a
0.1 mL
Primary
824.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit