Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain 5 Antibody (OTI7D2), DyLight 755, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270328IR

Catalog No. NBP270328IR


Add to Cart

Description

Description

Calpain 5 Monoclonal specifically detects Calpain 5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Calpain 5
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CAPN5
Recombinant protein fragment corresponding to amino acids 274-578 of human Calpain 5 (NP_004046) produced in E.coli.
0.1 mL
726.0
Store at 4°C in the dark.
Immunohistochemistry
OTI7D2
Immunohistochemistry
calpain 5, Calpain htra-3, calpain-5, EC 3.4.22, EC 3.4.22.54, FLJ46245, HTRA3NCL3, nCL-3EC 3.4.22.-, New calpain 3
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit