Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain 5 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI7D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270328AF405

Catalog No. N270328A405


Add to cart

Description

Description

Calpain 5 Monoclonal antibody specifically detects Calpain 5 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Calpain 5
OTI7D2
Immunohistochemistry
Affinity Purified, Conjugated
CAPN5
Recombinant protein fragment corresponding to amino acids 274-578 of human Calpain 5 (NP_004046) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
calpain 5, Calpain htra-3, calpain-5, EC 3.4.22, EC 3.4.22.54, FLJ46245, HTRA3NCL3, nCL-3EC 3.4.22.-, New calpain 3
Mouse
IgG1
0.1 mL
Primary
726.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit