Learn More
Alfa Aesar™ Calpain Inhibitor X
Manufacturer:  Alfa Aesar™ J65826LB0
 View more versions of this product
Catalog No. AAJ65826LB0
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
5mg
2°C to 8°C
Calpain Inhibitor X
C21H31N3O5
Z-Leu-α-aminobutyric acid-CONHC2H5; Z-Leu-Abu-CONHC2H5
White to off-white
Solid
A cell-permeable dipeptidyl →-ketoamide that acts as a potent, reversible, and active site inhibitor of calpain-1 and -2.
405.50
Light Sensitive