Learn More
Alfa Aesar™ Calpain Inhibitor XII
Manufacturer:  Alfa Aesar™ J64751LB0
 View more versions of this product
Catalog No. AAJ64751LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
5mg
2°C to 8°C
Calpain Inhibitor
C26H34N4O5
Z-L-Nva-CONH-CH2-2-Py
Pale yellow
Solid
A cell-permeable dipeptidyl →-ketoamide compound that acts as a potent, selective, reversible, and active site inhibitor of calpain-1
482.60