Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpastatin Antibody (OTI1E7), PE, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270338PE

Catalog No. NBP270338PE


Add to Cart

Description

Description

Calpastatin Monoclonal specifically detects Calpastatin in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Calpastatin
Monoclonal
PE
PBS with 0.05% Sodium Azide
CAST
Full length recombinant protein of human Calpastatin (NP_001741) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Flow Cytometry
OTI1E7
Flow Cytometry
BS-17, Calpain inhibitor, calpastatin, MGC9402, Sperm BS-17 component
Mouse
Affinity Purified
Hypoxia, Neuroscience
831.0
Store at 4°C in the dark.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit