Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpastatin Antibody (OTI1E7), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270338AF700
Description
Calpastatin Monoclonal specifically detects Calpastatin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Calpastatin | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CAST | |
Full length recombinant protein of human Calpastatin (NP_001741) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1E7 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
BS-17, Calpain inhibitor, calpastatin, MGC9402, Sperm BS-17 component | |
Mouse | |
Affinity Purified | |
Hypoxia, Neuroscience | |
831.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction