Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpastatin Mouse anti-Human, PE-Cyanine7, Clone: OTI1E7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270338PECY7

Catalog No. N270338PEC7


Add to cart

Description

Description

Calpastatin Monoclonal antibody specifically detects Calpastatin in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Calpastatin
OTI1E7
Flow Cytometry
Affinity Purified, Conjugated
CAST
Full length recombinant protein of human Calpastatin (NP_001741) produced in HEK293T cells.
Immunogen affinity purified
Hypoxia, Neuroscience
Primary
831.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
BS-17, Calpain inhibitor, calpastatin, MGC9402, Sperm BS-17 component
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit