Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpastatin Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Calpastatin
Applications KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094671
View Documents
Novus Biologicals
NBP303007100UL
100 μg Each for $320.71
Add to cart
 
NB094672
View Documents
Novus Biologicals
NBP30300720UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Calpastatin Polyclonal antibody specifically detects Calpastatin in Human, Mouse samples. It is validated for Western Blot, KnockDown
Specifications

Specifications

Calpastatin
Unconjugated
Rabbit
Hypoxia, Neuroscience
PBS with 50% glycerol, pH7.3.
831
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
KnockDown, Western Blot
Purified
RUO
Human, Mouse
BS-17, Calpain inhibitor, calpastatin, MGC9402, Sperm BS-17 component
A synthetic peptide corresponding to a sequence within amino acids 550-650 of human Calpastatin (NP_001177371.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit