Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CalPlus™ (Brand of Calcium Chloride Dihydrate), Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle

Chemical Identifiers

10035-04-8
CaCl2·2H2O

Suitable for Use in Hemodialysis

Formula Weight 147.01
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 0.84kg
Quantity 500g
Chemical Name or Material CalPlus™ (Brand of Calcium Chloride Dihydrate)
Grade Multi-Compendial/USP