Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calponin 1 Mouse anti-Human, Rat, Clone: rCNN1/832- Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
$603.00
Specifications
Antigen | Calponin 1 |
---|---|
Clone | rCNN1/832 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254381100
|
Novus Biologicals
NBP254381100UG |
100 μg |
Each for $603.00
|
|
|||||
NB254381200
|
Novus Biologicals
NBP254381200UG |
200 μg | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
Calponin 1 Monoclonal specifically detects Calponin 1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
Calponin 1 | |
1 mg/mL | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Cancer, Cytoskeleton Markers | |
PBS with No Preservative | |
1264 | |
Recombinant full-length human CNN1 protein | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
rCNN1/832 | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, smooth muscle, calponins, basic, Sm-Calp, SMCCcalponin-1 | |
CNN1 | |
IgG1 κ | |
Protein A or G purified | |
34 kDa |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title