Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calponin 2 Antibody (OTI2B5), FITC, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270421F

Catalog No. NBP270421F


Add to Cart

Description

Description

Calponin 2 Monoclonal specifically detects Calponin 2 in Human, Mouse, Rat, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Calponin 2
Monoclonal
FITC
PBS with 0.05% Sodium Azide
CNN2
Full length recombinant protein of human Calponin 2 (NP_004359) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat, Monkey
IgG1
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI2B5
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
calponin 2, Calponin H2, smooth muscle, calponin-2, Neutral calponin
Mouse
Affinity Purified
Signal Transduction
1265.0
Store at 4°C in the dark.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit