Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calponin 2 Mouse anti-Human, Primate, Clone: OTI2B5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270421

Catalog No. NBP270421


Add to cart

Description

Description

Calponin 2 Monoclonal antibody specifically detects Calponin 2 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Calponin 2
OTI2B5
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
calponin 2, Calponin H2, smooth muscle, calponin-2, Neutral calponin
Mouse
IgG1
Immunogen affinity purified
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
LYOPH
Western Blot 1:500-2000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100
Affinity Purified, Lyophilized
CNN2
Full length human recombinant protein of human CNN2(NP_004359) produced in HEK293T cell.
33.5 kDa
100 μg
Primary
1265
Human, Primate
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit