Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calprotectin Mouse anti-Human, Clone: CPT/1028, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14435

Catalog No. 89-034-518


Add to cart

Description

Description

Mouse monoclonal antibody raised against human calprotectin.

  • High specificity
  • High reactivity
Specifications

Specifications

Calprotectin
CPT/1028
Mouse monoclonal antibody raised against human calprotectin.
In 1 mM PBS (0.05% BSA and 0.05% azide).
Liquid
S100A8
Recombinant protein corresponding to human calprotectin.
Polyethylene Glycol (PEG) precipitation
RUO
Primary
6279|6280
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P05109; P06702
60B8AG|CAGA|CFAG|CGLA|CP-10|L1Ag|MA387|MIF|MRP8|NIF|P8
Mouse
IgM, kappa
100 ug
Store at 4°C.
Monoclonal
Recognizes the L1 or calprotectin molecule, an intra-cytoplasmic antigen comprising of a 12kDa alpha chain and a 14kDa beta chain. This antibody reacts with neutrophils, monocytes, macrophages, and squamous mucosal epithelia and is important for identifying macrophages in tissue sections.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)