Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CALR3 Mouse anti-Human, Clone: OTI1G4, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA505888
Description
Calreticulins, such as Calreticulin 3, are Ca(2+)-binding chaperones localized mainly in the endoplasmic/sarcoplasmic reticulum.Specifications
CALR3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96L12 | |
CMH19, CRT2, CT93 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
125972 |
Western Blot | |
OTI1G4 | |
Unconjugated | |
CALR3 | |
Liquid | |
CALR3 | |
Full length human recombit protein of human CALR3 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title